LIF947290594 Life Chemicals Inc.

2-(3-methoxy-3-oxopropyl)-1,1,1-trimethylhydrazinium bromide

  • MDL number MFCD10465251
  • Cas Number : 106966-25-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart