LIF947290599 Life Chemicals Inc.

1-(benzo[d]oxazol-2-yl)piperidin-4-ol

  • MDL number MFCD10414187
  • Cas Number : 1015946-14-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart