LIF947290626 Life Chemicals Inc.

2-[(3-fluorobenzyl)thio]-4,5-dihydro-1H-imidazole hydrobromide

  • MDL number MFCD16661056
  • Cas Number : 1351589-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart