LIF947290632 Life Chemicals Inc.

3-amino-1-(1-naphthyl)propan-1-ol

  • MDL number MFCD21177781
  • Cas Number : 861092-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart