LIF947290634 Life Chemicals Inc.

2-[(2-chloro-6-fluorobenzyl)thio]-4,5-dihydro-1H-imidazole hydrochloride

  • MDL number MFCD06480364
  • Cas Number : 1094032-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart