LIF947290638 Life Chemicals Inc.

2-(piperidin-4-yloxy)nicotinonitrile hydrochloride

  • MDL number MFCD09877665
  • Cas Number : 1185310-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart