LIF947290655 Life Chemicals Inc.

2-(2-thienyl)piperazine dihydrochloride

  • MDL number MFCD22587840
  • Cas Number : 1351590-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart