LIF947290656 Life Chemicals Inc.

6-methyl-4-(pyrrolidin-3-yloxy)-2H-pyran-2-one hydrochloride

  • MDL number MFCD28017006
  • Cas Number : 1864056-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart