LIF947290659 Life Chemicals Inc.

methyl amino(4-methylphenyl)acetate hydrochloride

  • MDL number MFCD08456676
  • Cas Number : 134722-09-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart