LIF947290675 Life Chemicals Inc.

2-amino-1-(1-naphthyl)ethanol

  • MDL number MFCD09041641
  • Cas Number : 23913-55-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart