LIF947290710 Life Chemicals Inc.

2-(2-chlorophenyl)-2-methoxyethan-1-amine

  • MDL number MFCD16062870
  • Cas Number : 24065-45-0
  • Molecular Weight : 185.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart