LIF947290715 Life Chemicals Inc.

4-(1,3-thiazol-2-yloxy)benzoic acid

  • MDL number MFCD09753043
  • Cas Number : 56355-20-5
  • Molecular Weight : 221.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart