LIF947290718 Life Chemicals Inc.

1,3-oxazolidine-2,4-dione

  • MDL number MFCD02691902
  • Cas Number : 2346-26-1
  • Molecular Weight : 101.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart