LIF947290759 Life Chemicals Inc.

5,6-dimethylpyridazin-3-ol

  • MDL number MFCD16871377
  • Cas Number : 100114-19-0
  • Molecular Weight : 124.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart