LIF947290763 Life Chemicals Inc.

5-(benzyloxy)-4-chloropyrimidine

  • MDL number MFCD23701731
  • Cas Number : 91063-23-9
  • Molecular Weight : 220.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart