LIF947290764 Life Chemicals Inc.

2-(1,1-dioxido-1,2,5-thiadiazolidin-2-yl)acetic acid

  • MDL number MFCD21997197
  • Cas Number : 1368437-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart