LIF947290765 Life Chemicals Inc.

tert-butyl 4-ethynyl-2,2-dimethyloxazolidine-3-carboxylate

  • MDL number MFCD16620634
  • Cas Number : 1087043-97-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart