LIF947290767 Life Chemicals Inc.

2-chloro-4-(1H-tetrazol-5-yl)pyridine

  • MDL number MFCD10696104
  • Cas Number : 1196152-14-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart