LIF947290774 Life Chemicals Inc.

[2,4'-bipyridine]-4-carboxylic acid

  • MDL number MFCD15476023
  • Cas Number : 1214342-58-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart