LIF947290792 Life Chemicals Inc.

2-[6-oxo-3-(thiophen-2-yl)-1,6-dihydropyridazin-1-yl]propanoic acid

  • MDL number MFCD09881889
  • Cas Number : 1017378-02-7
  • Molecular Weight : 250.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart