LIF947290795 Life Chemicals Inc.

2-cyclobutyl-2-methylpropanoic acid

  • MDL number MFCD27384462
  • Cas Number : 1603359-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart