LIF947290796 Life Chemicals Inc.

{1-[(thiophen-3-yl)methyl]piperidin-4-yl}methanol

  • MDL number MFCD16162920
  • Cas Number : 1249314-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart