LIF947290797 Life Chemicals Inc.

[1-(2,3-dimethylphenyl)-1H-1,2,3-triazol-4-yl]methanol

  • MDL number MFCD16747671
  • Cas Number : 1251237-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart