LIF947290805 Life Chemicals Inc.

1-[(1,4-diazepan-1-yl)methyl]cyclopentan-1-ol

  • MDL number MFCD21182513
  • Cas Number : 1492280-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart