LIF947290820 Life Chemicals Inc.

1-[2-(4-chlorophenyl)ethyl]-1H-1,3-benzodiazol-5-amine

  • MDL number MFCD17085404
  • Cas Number : 1493318-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart