LIF947290821 Life Chemicals Inc.

(4S)-1,2-oxazolidin-4-ol hydrochloride

  • MDL number MFCD20483793
  • Cas Number : 338464-55-4
  • Molecular Weight : 125.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart