LIF947290834 Life Chemicals Inc.

1-[(1,4-diazepan-1-yl)methyl]cyclobutan-1-ol

  • MDL number MFCD30695162
  • Cas Number : 2098500-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart