LIF947290836 Life Chemicals Inc.

4-[(1-hydroxycyclobutyl)methyl]piperazin-2-one

  • MDL number MFCD30830323
  • Cas Number : 2098500-76-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart