LIF948329735 Life Chemicals Inc.

2-(4,4-difluorocyclohexyl)acetic acid

  • MDL number MFCD11042913
  • Cas Number : 915030-40-9
  • Molecular Weight : 178.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart