LIF948329809 Life Chemicals Inc.

4-methylbenzene-1-sulfonic acid; 6-oxa-3-azabicyclo[3.1.1]heptane

  • MDL number MFCD27665413
  • Cas Number : 1339953-58-0
  • Molecular Weight : 271.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart