LIF948329825 Life Chemicals Inc.

ethyl 3-oxocyclopentane-1-carboxylate

  • MDL number MFCD01320352
  • Cas Number : 5400-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart