LIF948329836 Life Chemicals Inc.

(2-methylpyrimidin-5-yl)methanol

  • MDL number MFCD09414732
  • Cas Number : 2239-83-0
  • Molecular Weight : 124.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart