LIFH03B69E00 Life Chemicals Inc.

4-(piperidin-4-yloxy)benzamide

  • MDL number MFCD08061080
  • Cas Number : 609781-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart