LIFH03B69E05 Life Chemicals Inc.

4-(2,2-difluorocyclopropyl)piperidine hydrochloride

  • MDL number MFCD30004411
  • Cas Number : 2097991-05-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart