LIFH03B69E1E Life Chemicals Inc.

METHYL TRANS-4-(TRIFLUOROMETHYL)PYRROLIDINE-3-CARBOXYLATE HYDROCHLORIDE

  • MDL number MFCD09910398
  • Cas Number : 1212115-75-7
  • Molecular Weight : 233.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart