LIFH03B69E2C Life Chemicals Inc.

2-(4-methylphenyl)-2H-1,2,3,4-tetrazole-5-carboxylic acid

  • MDL number MFCD21882987
  • Cas Number : 65251-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart