LIFH03B69E2E Life Chemicals Inc.

2-amino-N-(pyridin-3-yl)benzamide

  • MDL number MFCD03015406
  • Cas Number : 76102-92-6
  • Molecular Weight : 213.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart