LIFH03B69E5C Life Chemicals Inc.

5,7-dichloro-1,2,3,4-tetrahydronaphthalen-1-one

  • MDL number MFCD08544415
  • Cas Number : 248607-58-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart