LIFH03B69E5D Life Chemicals Inc.

6,8-dichloro-1,2,3,4-tetrahydronaphthalen-1-one

  • MDL number MFCD18208420
  • Cas Number : 1260017-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart