LIFH03CD7582 Life Chemicals Inc.

2-(2,3-dimethylphenoxy)propanoic acid

  • MDL number MFCD03419315
  • Cas Number : 22504-84-3
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart