LIFH03DC484D Life Chemicals Inc.

N-[(4-methoxyphenyl)methyl]cyclopropanamine

  • MDL number MFCD07310991
  • Cas Number : 70894-71-2
  • Molecular Weight : 177.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart