LIFH03DC4879 Life Chemicals Inc.

4,5,6-trimethylpyridine-3-carbonitrile

  • MDL number MFCD26381824
  • Cas Number : 847154-74-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart