LIFH03DC4880 Life Chemicals Inc.

1-(2-methylpropyl)-decahydropyrido[4,3-e][1,4]oxazepin-2-one hydrochloride

  • Cas Number : 2320340-16-5
  • Molecular Weight : 262.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart