LIFH03DC4881 Life Chemicals Inc.

1-(2-methoxyethyl)-decahydropyrido[4,3-e][1,4]oxazepin-2-one hydrochloride

  • Cas Number : 2320224-87-9
  • Molecular Weight : 264.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart