LIFH03DC488B Life Chemicals Inc.

2-(4-benzoylphenoxy)acetic acid

  • MDL number MFCD02725448
  • Cas Number : 6322-83-4
  • Molecular Weight : 256.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart