LIFH03DC48AE Life Chemicals Inc.

trans-4-(propan-2-yloxy)oxolan-3-ol

  • MDL number MFCD31619603
  • Cas Number : 2377004-16-3
  • Molecular Weight : 146.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart