LIFH03DC48AF Life Chemicals Inc.

1-(4-methylphenoxy)propan-2-amine

  • MDL number MFCD06248348
  • Cas Number : 61711-87-3
  • Molecular Weight : 165.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart