LIFH04830473 Life Chemicals Inc.

O-chlorophenoxyacetic acid

  • MDL number MFCD00004298
  • Cas Number : 614-61-9
  • Molecular Weight : 186.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart