LIFH04830495 Life Chemicals Inc.

(E)-N-[(2,6-dimethoxyphenyl)methylidene]hydroxylamine

  • MDL number MFCD02683867
  • Cas Number : 174966-94-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart