LIFH0C9551CA Life Chemicals Inc.

1-aminobutan-2-one hydrochloride

  • MDL number MFCD13152262
  • Cas Number : 108661-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart